3-Brom-4-nitroanilin

Lieferant: ACTIVATE SCIENTIFIC

AS33812G1 AS33812G10 AS33812G5
ACTIAS33812G1EA 0 CHF
ACTIAS33812G1 ACTIAS33812G10 ACTIAS33812G5
3-Brom-4-nitroanilin
3-Brom-4-nitroanilin
Formel: C₆H₅BrN₂O₂
MW: 217.0200 g/mol
MDL Number: MFCD07783047
CAS-Nummer: 40787-96-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR