2-Brom-3-fluorphenol

Lieferant: ACTIVATE SCIENTIFIC

AS22843G1 AS22843G25 AS22843G5
ACTIAS22843G1EA 0 CHF
ACTIAS22843G1 ACTIAS22843G25 ACTIAS22843G5
2-Brom-3-fluorphenol
2-Brom-3-fluorphenol
Formel: C₆H₄BrFO
MDL Number: MFCD04037220
CAS-Nummer: 443-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR