2-Brom-3,4-difluoranilin

Lieferant: ACTIVATE SCIENTIFIC

AS32895G1 AS32895G5
ACTIAS32895G1EA 0 CHF
ACTIAS32895G1 ACTIAS32895G5
2-Brom-3,4-difluoranilin
2-Brom-3,4-difluoranilin
Formel: C₆H₄BrF₂N
MW: 208,00 g/mol
MDL Number: MFCD11035924
CAS-Nummer: 1092349-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR