2-Brom-6-chlorbenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS35708G1 AS35708G5
ACTIAS35708G1EA 0 CHF
ACTIAS35708G1 ACTIAS35708G5
2-Brom-6-chlorbenzaldehyd
2-Bromo-6-chlorobenzaldehyde
Formel: C₇H₄BrClO
MW: 219,46 g/mol
MDL Number: MFCD00266792
CAS-Nummer: 64622-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR