2-Brom-3-chloranilin

Lieferant: ACTIVATE SCIENTIFIC

AS33038G1 AS33038G5
ACTIAS33038G1EA 0 CHF
ACTIAS33038G1 ACTIAS33038G5
2-Brom-3-chloranilin
2-Brom-3-chloranilin
Formel: C₆H₅BrClN
MDL Number: MFCD10699543
CAS-Nummer: 96558-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR