6-Bromanthranilsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Amino-6-bromobenzoic acid

AS22050G1 AS22050G25 AS22050G5
ACTIAS22050G1EA 18 CHF
ACTIAS22050G1 ACTIAS22050G25 ACTIAS22050G5
6-Bromanthranilsäure
6-Bromoanthranilic acid
Formel: C₇H₆BrNO₂
MW: 216,03 g/mol
MDL Number: MFCD03618455
CAS-Nummer: 20776-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR