4-Amino-2,5-dimethylphenol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Amino-2,5-dimethylphenol

AS37542G10 AS37542G5
ACTIAS37542G10EA 0 CHF
ACTIAS37542G10 ACTIAS37542G5
4-Amino-2,5-dimethylphenol
4-Amino-2,5-xylenol
Formel: C₈H₁₁NO
MW: 137,1800 g/mol
MDL Number: MFCD00007873
CAS-Nummer: 3096-71-7
EINECS: 221-449-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR