3-Methylisonicotinsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Methyl-4-pyridinecarboxylic acid

AS1184G1 AS1184G5
ACTIAS1184G1EA 0 CHF
ACTIAS1184G1 ACTIAS1184G5
3-Methylisonicotinsäure
3-Methylisonicotinsäure
Formel: C₇H₇NO₂
MDL Number: MFCD00128870
CAS-Nummer: 4021-12-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR