5-p-Tolylpyridin-2-ylamin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-(p-Tolyl)pyridin-2-amine, 5-(4-Methylphenyl)pyridin-2-amine

AS30302G1 AS30302G5
ACTIAS30302G1EA 0 CHF
ACTIAS30302G1 ACTIAS30302G5
5-p-Tolylpyridin-2-ylamin
5-p-Tolylpyridin-2-ylamin
Formel: C₁₂H₁₂N₂
MW: 184,240 g/mol
MDL Number: MFCD06797790
CAS-Nummer: 503536-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR