4-Brom-3-(trifluormethyl)benzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS35778G1 AS35778G5
ACTIAS35778G1EA 0 CHF
ACTIAS35778G1 ACTIAS35778G5
4-Brom-3-(trifluormethyl)benzaldehyd
4-Bromo-3-(trifluoromethyl)benzaldehyde
Formel: C₈H₄BrF₃O
MDL Number: MFCD08059130
CAS-Nummer: 34328-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR