2-Brom-4-(trifluormethyl)benzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS32594G1 AS32594G5
ACTIAS32594G1EA 0 CHF
ACTIAS32594G1 ACTIAS32594G5
2-Brom-4-(trifluormethyl)benzaldehyd
2-Bromo-4-(trifluoromethyl)benzaldehyde
Formel: C₈H₄BrF₃O
MW: 253.0200 g/mol
MDL Number: MFCD09835101
CAS-Nummer: 85118-24-7
EINECS: 285-677-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR