4'-Benzyloxy-2'-hydroxyacetophenon

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS35948G1EA 0 CHF
ACTIAS35948G1 ACTIAS35948G5
4'-Benzyloxy-2'-hydroxyacetophenon
4'-Benzyloxy-2'-hydroxyacetophenon
Formel: C₁₅H₁₄O₃
MW: 242,2699 g/mol
MDL Number: MFCD00054612
CAS-Nummer: 29682-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR