4-Nitro-2,3-xylidin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2,3-Dimethyl-4-nitroaniline

AS35787G1 AS35787G5
ACTIAS35787G1EA 0 CHF
ACTIAS35787G1 ACTIAS35787G5
4-Nitro-2,3-xylidin
4-Nitro-2,3-xylidin
Formel: C₈H₁₀N₂O₂
MW: 166,1772 g/mol
MDL Number: MFCD00130024
CAS-Nummer: 80879-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR