5-Nitro-m-toluidin

Lieferant: ACTIVATE SCIENTIFIC
Gefahr

Synonyms: 3-Methyl-5-nitroaniline

AS32433G1 AS32433G5
ACTIAS32433G1EA 0 CHF
ACTIAS32433G1 ACTIAS32433G5
5-Nitro-m-toluidin
5-Nitro-m-toluidin
Formel: C₇H₈N₂O₂
MW: 152,1500 g/mol
MDL Number: MFCD00082655
CAS-Nummer: 618-61-1
UN: 2660
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR