2-Nitro-m-toluidin
Lieferant: ACTIVATE SCIENTIFIC
|
Gefahr
|
Synonyms:
3-Methyl-2-nitroaniline
AS33855G1
AS33855G10
AS33855G5
ACTIAS33855G1EA
0
CHF
ACTIAS33855G1
ACTIAS33855G10
ACTIAS33855G5
2-Nitro-m-toluidin
2-Nitro-m-toluidin
Formel:
C₇H₈N₂O₂ MW: 152,1500 g/mol |
MDL Number:
MFCD00074851 CAS-Nummer: 601-87-6 UN: 2660 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...