4,4-Difluorcyclohexanol

Lieferant: ACTIVATE SCIENTIFIC

AS32321G1 AS32321G5
ACTIAS32321G1EA 0 CHF
ACTIAS32321G1 ACTIAS32321G5
4,4-Difluorcyclohexanol
4,4-Difluorcyclohexanol
Formel: C₆H₁₀F₂O
MW: 136,1398 g/mol
MDL Number: MFCD10000566
CAS-Nummer: 22419-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR