3-Amino-4-fluorphenol

Lieferant: ACTIVATE SCIENTIFIC

AS31331G1 AS31331G5
ACTIAS31331G1EA 0 CHF
ACTIAS31331G1 ACTIAS31331G5
3-Amino-4-fluorphenol
3-Amino-4-fluorphenol
Formel: C₆H₆FNO
MW: 127,1200 g/mol
MDL Number: MFCD09033850
CAS-Nummer: 62257-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR