2-Amino-5-fluor-6-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS32524G1 AS32524G5
ACTIAS32524G1EA 0 CHF
ACTIAS32524G1 ACTIAS32524G5
2-Amino-5-fluor-6-methylpyridin
2-Amino-5-fluoro-6-methylpyridine
Formel: C₆H₇FN₂
MW: 126,13 g/mol
MDL Number: MFCD09835298
CAS-Nummer: 110919-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR