3-Amino-5-fluorpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Fluoropyridin-3-amine

AS31204G1 AS31204G5
ACTIAS31204G1EA 0 CHF
ACTIAS31204G1 ACTIAS31204G5
3-Amino-5-fluorpyridin
3-Amino-5-fluoropyridine
Formel: C₅H₅FN₂
MW: 112,11 g/mol
MDL Number: MFCD00234073
CAS-Nummer: 210169-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR