2,5-Difluor-p-toluylsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2,5-Difluoro-4-methylbenzoic acid

AS33675G1 AS33675G5
ACTIAS33675G1EA 0 CHF
ACTIAS33675G1 ACTIAS33675G5
2,5-Difluor-p-toluylsäure
2,5-Difluor-p-toluylsäure
Formel: C₈H₆F₂O₂
MW: 172,13 g/mol
MDL Number: MFCD08063230
CAS-Nummer: 103877-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR