4,6-Difluor-m-toluylsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2,4-Difluoro-5-methylbenzoic acid

AS33793G1 AS33793G5
ACTIAS33793G1EA 0 CHF
ACTIAS33793G1 ACTIAS33793G5
4,6-Difluor-m-toluylsäure
4,6-Difluor-m-toluylsäure
Formel: C₈H₆F₂O₂
MW: 172,13 g/mol
MDL Number: MFCD09835285
CAS-Nummer: 367954-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR