2,4-Difluor-m-toluylsäure

Lieferant: ACTIVATE SCIENTIFIC

AS32229G1 AS32229G5
ACTIAS32229G1EA 0 CHF
ACTIAS32229G1 ACTIAS32229G5
2,4-Difluor-m-toluylsäure
2,4-Difluor-m-toluylsäure
Formel: C₈H₆F₂O₂
MDL Number: MFCD03265223
CAS-Nummer: 112857-68-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR