5-Brom-m-kresol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Bromo-5-methylphenol

AS33622G1 AS33622G10 AS33622G5
ACTIAS33622G1EA 0 CHF
ACTIAS33622G1 ACTIAS33622G10 ACTIAS33622G5
5-Brom-m-kresol
5-Brom-m-kresol
Formel: C₇H₇BrO
MW: 187,0300 g/mol
MDL Number: MFCD09038281
CAS-Nummer: 74204-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR