3-Amino-4-bromphenol

Lieferant: ACTIVATE SCIENTIFIC

AS34135G1 AS34135G10 AS34135G5
ACTIAS34135G1EA 0 CHF
ACTIAS34135G1 ACTIAS34135G10 ACTIAS34135G5
3-Amino-4-bromphenol
3-Amino-4-bromphenol
Formel: C₆H₆BrNO
MW: 188,0200 g/mol
MDL Number: MFCD09260886
CAS-Nummer: 100367-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR