3-Propoxyphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

AS51528G1 AS51528G5
ACTIAS51528G1EA 0 CHF
ACTIAS51528G1 ACTIAS51528G5
3-Propoxyphenylboronsäure
3-Propoxyphenylboronsäure
Formel: C₉H₁₃BO₃
MW: 180,0100 g/mol
MDL Number: MFCD03427190
CAS-Nummer: 149557-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR