3-Methoxy-o-toluidin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
3-Methoxy-2-methylaniline
AS22046G25
AS22046G5
ACTIAS22046G25EA
195
CHF
ACTIAS22046G25
ACTIAS22046G5
3-Methoxy-o-toluidin
3-Methoxy-o-toluidin
Formel:
C₈H₁₁NO MW: 137,1800 g/mol |
MDL Number:
MFCD06412568 CAS-Nummer: 19500-02-8 |
Learn more
![](/m-ch.vwr.com/de_CH/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...