4-Hydroxy-3-methylbenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS34247G1 AS34247G5
ACTIAS34247G1EA 0 CHF
ACTIAS34247G1 ACTIAS34247G5
4-Hydroxy-3-methylbenzaldehyd
4-Hydroxy-3-methylbenzaldehyde
Formel: C₈H₈O₂
MW: 136,1479 g/mol
MDL Number: MFCD00012360
CAS-Nummer: 15174-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR