4-Hydroxy-2-methylbenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS30957G1 AS30957G5
ACTIAS30957G1EA 0 CHF
ACTIAS30957G1 ACTIAS30957G5
4-Hydroxy-2-methylbenzaldehyd
4-Hydroxy-2-methylbenzaldehyde
Formel: C₈H₈O₂
MW: 136,1479 g/mol
MDL Number: MFCD04037443
CAS-Nummer: 41438-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR