4-Methylsalicylaldehyd

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Hydroxy-4-methylbenzaldehyde

AS35386G1 AS35386G5
ACTIAS35386G1EA 0 CHF
ACTIAS35386G1 ACTIAS35386G5
4-Methylsalicylaldehyd
4-Methylsalicylaldehyd
Formel: C₈H₈O₂
MW: 136,15 g/mol
MDL Number: MFCD00799550
CAS-Nummer: 698-27-1
EINECS: 211-813-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR