4-Brom-5-chloranthranilsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2-Amino-4-bromo-5-chlorobenzoic acid
AS33709G1
AS33709G5
ACTIAS33709G1EA
0
CHF
ACTIAS33709G1
ACTIAS33709G5
4-Brom-5-chloranthranilsäure
4-Brom-5-chloranthranilsäure
Formel:
C₇H₅BrClNO₂ MW: 250,4800 g/mol |
MDL Number:
MFCD10000554 CAS-Nummer: 150812-32-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...