3-Brombenzaldehydethylenacetal
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2-(3-Bromophenyl)-1,3-dioxolane
AS21809G100
AS21809G25
ACTIAS21809G100EA
0
CHF
ACTIAS21809G100
ACTIAS21809G25
3-Brombenzaldehydethylenacetal
3-Brombenzaldehydethylenacetal
Formel:
C₉H₉BrO₂ MW: 229.07 g/mol |
MDL Number:
MFCD00003209 CAS-Nummer: 17789-14-9 EINECS: 241-766-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...