2-Thiophenboronsäurepinakolester

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS20674G25EA 0 CHF
ACTIAS20674G25 ACTIAS20674G5
2-Thiophenboronsäurepinakolester
2-Thiophenboronsäurepinakolester
Formel: C₁₀H₁₅BO₂S
MW: 210,1 g/mol
MDL Number: MFCD05663878
CAS-Nummer: 193978-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR