6-(3-Fluorphenyl)-2-pyridincarbonsäure

Lieferant: ACTIVATE SCIENTIFIC

AS24608G1 AS24608G5
ACTIAS24608G1EA 367 CHF
ACTIAS24608G1 ACTIAS24608G5
6-(3-Fluorphenyl)-2-pyridincarbonsäure
6-(3-Fluorophenyl)-2-pyridinecarboxylic acid
Formel: C₁₂H₈FNO₂
MDL Number: MFCD06410053
CAS-Nummer: 887982-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR