5-(4-Fluorphenyl)-3-pyridincarbaldehyd

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-(4-Fluorophenyl)nicotinaldehyde

AS24610G1 AS24610G5
ACTIAS24610G1EA 0 CHF
ACTIAS24610G1 ACTIAS24610G5
5-(4-Fluorphenyl)-3-pyridincarbaldehyd
5-(4-Fluorphenyl)-3-pyridincarbaldehyd
Formel: C₁₂H₈FNO
MDL Number: MFCD02093924
CAS-Nummer: 381684-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR