6-(3,5-Difluorphenyl)-3-pyridincarbaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS24488G1 AS24488G5
ACTIAS24488G1EA 0 CHF
ACTIAS24488G1 ACTIAS24488G5
6-(3,5-Difluorphenyl)-3-pyridincarbaldehyd
6-(3,5-Difluorphenyl)-3-pyridincarbaldehyd
Formel: C₁₂H₇F₂NO
MDL Number: MFCD04115470
CAS-Nummer: 898404-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR