6-(3,4-Difluorphenyl)-3-pyridincarbaldehyd
Lieferant: ACTIVATE SCIENTIFIC
AS24632G1
AS24632G5
ACTIAS24632G1EA
0
CHF
ACTIAS24632G1
ACTIAS24632G5
6-(3,4-Difluorphenyl)-3-pyridincarbaldehyd
6-(3,4-Difluorphenyl)-3-pyridincarbaldehyd
Formel:
C₁₂H₇F₂NO |
CAS-Nummer:
898404-54-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...