2-Methyl-6-(trifluormethyl)nicotinsäure

Lieferant: ACTIVATE SCIENTIFIC

AS36513G1 AS36513G5
ACTIAS36513G1EA 0 CHF
ACTIAS36513G1 ACTIAS36513G5
2-Methyl-6-(trifluormethyl)nicotinsäure
2-Methyl-6-(trifluormethyl)nicotinsäure
Formel: C₈H₆F₃NO₂
MDL Number: MFCD01311987
CAS-Nummer: 261635-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR