2-Methyl-6-(trifluormethyl)nicotinsäurehydrazid

Lieferant: ACTIVATE SCIENTIFIC

AS23645G1 AS23645G5
ACTIAS23645G1EA 0 CHF
ACTIAS23645G1 ACTIAS23645G5
2-Methyl-6-(trifluormethyl)nicotinsäurehydrazid
2-Methyl-6-(trifluormethyl)nicotinsäurehydrazid
Formel: C₈H₈F₃N₃O
MDL Number: MFCD01565843
CAS-Nummer: 402479-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR