6-(Trifluormethyl)nicotinoylchlorid

Lieferant: ACTIVATE SCIENTIFIC

AS20514G1 AS20514G5
ACTIAS20514G1EA 89 CHF
ACTIAS20514G1 ACTIAS20514G5
6-(Trifluormethyl)nicotinoylchlorid
6-(Trifluormethyl)nicotinoylchlorid
Formel: C₇H₃ClF₃NO
MW: 209,5530 g/mol
MDL Number: MFCD01862645
CAS-Nummer: 358780-13-9
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR