4-(Trifluormethyl)nicotinonitril

Lieferant: ACTIVATE SCIENTIFIC

AS32610G1 AS32610G5
ACTIAS32610G1EA 0 CHF
ACTIAS32610G1 ACTIAS32610G5
4-(Trifluormethyl)nicotinonitril
4-(Trifluormethyl)nicotinonitril
Formel: C₇H₃F₃N₂
MDL Number: MFCD00153107
CAS-Nummer: 13600-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR