2-Amino-5-fluor-3-iodpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS30993G1 AS30993G5
ACTIAS30993G1EA 0 CHF
ACTIAS30993G1 ACTIAS30993G5
2-Amino-5-fluor-3-iodpyridin
2-Amino-5-fluoro-3-iodopyridine
Formel: C₅H₄FIN₂
MW: 238,0015 g/mol
MDL Number: MFCD08688587
CAS-Nummer: 823218-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR