5-Methoxyisatoic anhydride

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 6-Methoxy-1H-benzo[d][1,3]oxazine-2,4-dione, 6-Methoxy-2H-3,1-benzoxazine-2,4(1H)-dione

AS35389G1 AS35389G5
ACTIAS35389G1EA 0 CHF
ACTIAS35389G1 ACTIAS35389G5
5-Methoxyisatoic anhydride
5-Methoxyisatoic anhydride
Formel: C₉H₇NO₄
MW: 193,156 g/mol
MDL Number: MFCD02179411
CAS-Nummer: 37795-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR