2-Amino-4'-fluorbenzophenon

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (2-Aminophenyl)(4-fluorophenyl)methanone

AS24099G100 AS24099G25
ACTIAS24099G100EA 0 CHF
ACTIAS24099G100 ACTIAS24099G25
2-Amino-4'-fluorbenzophenon
2-Amino-4'-fluorbenzophenon
Formel: C₁₃H₁₀FNO
MDL Number: MFCD06658166
CAS-Nummer: 3800-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR