1-Iod-4-methylimidazoliumchlorid

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Iodo-1-methyl-1H-imidazole hydrochloride

AS50085G1 AS50085G5
ACTIAS50085G1EA 0 CHF
ACTIAS50085G1 ACTIAS50085G5
1-Iod-4-methylimidazoliumchlorid
1-Iod-4-methylimidazoliumchlorid
Formel: C₄H₆ClIN₂
MDL Number: MFCD07783893
CAS-Nummer: 871333-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR