3-Fluor-4-formylphenylboronsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
3-Fluoro-4-formylbenzeneboronic acid
AS35023G1
AS35023G10
AS35023G5
ACTIAS35023G1EA
0
CHF
ACTIAS35023G1
ACTIAS35023G10
ACTIAS35023G5
3-Fluor-4-formylphenylboronsäure
3-Fluor-4-formylphenylboronsäure
Formel:
C₇H₆BFO₃ MW: 167,93 g/mol |
MDL Number:
MFCD02093051 CAS-Nummer: 248270-25-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...