2-Fluor-5-formylphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Fluoro-5-formylbenzeneboronic acid

AS50119G1 AS50119G5
ACTIAS50119G1EA 0 CHF
ACTIAS50119G1 ACTIAS50119G5
2-Fluor-5-formylphenylboronsäure
2-Fluor-5-formylphenylboronsäure
Formel: C₇H₆BFO₃
MW: 167,93 g/mol
MDL Number: MFCD03095100
CAS-Nummer: 352534-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR