2-Chlor-4-fluor-5-nitrophenol

Lieferant: ACTIVATE SCIENTIFIC

AS22322G25 AS22322G5
ACTIAS22322G25EA 0 CHF
ACTIAS22322G25 ACTIAS22322G5
2-Chlor-4-fluor-5-nitrophenol
2-Chlor-4-fluor-5-nitrophenol
Formel: C₆H₃ClFNO₃
MDL Number: MFCD02670258
CAS-Nummer: 84478-75-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR