3-Chlor-4-hydroxyphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (3-Chloro-4-hydroxyphenyl)boronic acid

AS35267G10 AS35267G5
ACTIAS35267G10EA 0 CHF
ACTIAS35267G10 ACTIAS35267G5
3-Chlor-4-hydroxyphenylboronsäure
3-Chlor-4-hydroxyphenylboronsäure
Formel: C₆H₆BClO₃
MW: 172,3700 g/mol
MDL Number: MFCD09258739
CAS-Nummer: 182344-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR