5-Chlor-2-formylphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

AS51158G1 AS51158G5
ACTIAS51158G1EA 0 CHF
ACTIAS51158G1 ACTIAS51158G5
5-Chlor-2-formylphenylboronsäure
5-Chlor-2-formylphenylboronsäure
Formel: C₇H₆BClO₃
MDL Number: MFCD03425955
CAS-Nummer: 870238-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR