2-Chlor-5-formylphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

AS51160G1 AS51160G5
ACTIAS51160G1EA 0 CHF
ACTIAS51160G1 ACTIAS51160G5
2-Chlor-5-formylphenylboronsäure
2-Chlor-5-formylphenylboronsäure
Formel: C₇H₆BClO₃
MDL Number: MFCD06656269
CAS-Nummer: 1150114-78-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR